Merck
  • The environmental obesogen bisphenol A increases macrophage self-renewal.
The environmental obesogen bisphenol A increases macrophage self-renewal.
Cell and tissue research (2019-04-24)
Grace Ampem, Alexandra Junginger, Haidong Yu, Lajos Balogh, Julianna Thuróczy, Marion E Schneider, Tamás Röszer
ABSTRACT
Self-renewal of macrophages is important for the healthy development and replenishment of tissue-resident macrophage pools. How this mechanism is controlled by endocrine signals is still largely unexplored. Here, we show that the endocrine disruptor bisphenol A (BPA) increases macrophage self-renewal. This effect was associated with phosphorylation of extracellular signal-regulated kinase (ERK) and a slight increase in the expression of liver X receptor alpha (LXRα). We found that LXRα inhibition induced, while LXRα activation impeded, macrophage self-renewal. LXRα signaling hence may protect from excessive macrophage expansion. Self-renewing macrophages, however, had negligible LXRα expression when compared with quiescent macrophages. Accordingly, tissue-resident macrophage pools, which are dominated by quiescent macrophages, were rich in LXRα-expressing macrophages. Overall, we show that BPA increases macrophage self-renewal and that this effect, at least in part, can be inhibited by increasing LXRα expression. Since BPA is accumulated in the adipose tissue, it has the potential to increase self-renewal of adipose tissue macrophages, leading to a condition that might negatively impact adipose tissue health.
Sigma-Aldrich
Hanks′ Balanced Salt solution, With sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
TRI Reagent®, For processing tissues, cells cultured in monolayer or cell pellets
Sigma-Aldrich
GW3965 hydrochloride, ≥98% (HPLC), powder
Supelco
Bisphenol A, ≥99%
Sigma-Aldrich
25-Hydroxycholesterol, ≥98%
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
SR9238, ≥98% (HPLC)