Merck
  • Interaction of Synaptosomal-Associated Protein 25 with Neutral Sphingomyelinase 2: Functional Impact on the Sphingomyelin Pathway.
Interaction of Synaptosomal-Associated Protein 25 with Neutral Sphingomyelinase 2: Functional Impact on the Sphingomyelin Pathway.
Neuroscience (2019-11-26)
Jong Hoon Won, Hyung Jun Jeon, Seok Kyun Kim, In Chul Shin, Ji Min Jang, Hae Chan Ha, Moon Jung Back, Dae Kyong Kim
ABSTRACT
Neurotransmitter release is mediated by ceramide, which is generated by sphingomyelin hydrolysis. In the present study, we examined whether synaptosomal-associated protein 25 (SNAP-25) is involved in ceramide production and exocytosis. Neutral sphingomyelinase 2 (nSMase2) was partially purified from bovine brain and we found that SNAP-25 was enriched in the nSMase2-containing fractions. In rat synaptosomes and PC12 cells, the immunoprecipitation pellet of anti-SNAP-25 antibody showed higher nSMase activity than the immunoprecipitation pellet of anti-nSMase2 antibody. In PC12 cells, SNAP-25 was colocalized with nSMase2. Transfection of SNAP-25 small interfering RNA (siRNA) significantly inhibited nSMase2 translocation to the plasma membrane. A23187-induced ceramide production was concomitantly reduced in SNAP-25 siRNA-transfected PC12 cells compared with that in scrambled siRNA-transfected cells. Moreover, transfection of SNAP-25 siRNA inhibited dopamine release, whereas addition of C6-ceramide to the siRNA-treated cells moderately reversed this inhibition. Additionally, nSMase2 inhibition reduced dopamine release. Collectively, our results indicate that SNAP-25 interacts with nSMase2 during ceramide production, which mediates exocytosis and neurotransmitter release.
Product Description
Avanti
C18 Glucosyl(Ī²) Ceramide (d18:1/18:0), Avanti Researchā„¢ - A Croda Brand