Merck
  • A cell type-selective apoptosis-inducing small molecule for the treatment of brain cancer.
A cell type-selective apoptosis-inducing small molecule for the treatment of brain cancer.
Proceedings of the National Academy of Sciences of the United States of America (2019-03-09)
Natasha C Lucki, Genaro R Villa, Naja Vergani, Michael J Bollong, Brittney A Beyer, Jae Wook Lee, Justin L Anglin, Stephan H Spangenberg, Emily N Chin, Amandeep Sharma, Kevin Johnson, Philipp N Sander, Perry Gordon, Stephen L Skirboll, Heiko Wurdak, Peter G Schultz, Paul S Mischel, Luke L Lairson
ABSTRACT
Glioblastoma multiforme (GBM; grade IV astrocytoma) is the most prevalent and aggressive form of primary brain cancer. A subpopulation of multipotent cells termed GBM cancer stem cells (CSCs) play a critical role in tumor initiation, tumor maintenance, metastasis, drug resistance, and recurrence following surgery. Here we report the identification of a small molecule, termed RIPGBM, from a cell-based chemical screen that selectively induces apoptosis in multiple primary patient-derived GBM CSC cultures. The cell type-dependent selectivity of this compound appears to arise at least in part from redox-dependent formation of a proapoptotic derivative, termed cRIPGBM, in GBM CSCs. cRIPGBM induces caspase 1-dependent apoptosis by binding to receptor-interacting protein kinase 2 (RIPK2) and acting as a molecular switch, which reduces the formation of a prosurvival RIPK2/TAK1 complex and increases the formation of a proapoptotic RIPK2/caspase 1 complex. In an orthotopic intracranial GBM CSC tumor xenograft mouse model, RIPGBM was found to significantly suppress tumor formation in vivo. Our chemical genetics-based approach has identified a drug candidate and a potential drug target that provide an approach to the development of treatments for this devastating disease.
Sigma-Aldrich
RIPGBM, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
cRIPGBM, ≥98% (HPLC)
Pricing and availability is not currently available.