Merck
  • Synthesis of highly substituted isoquinolone derivatives by nickel-catalyzed annulation of 2-halobenzamides with alkynes.
Synthesis of highly substituted isoquinolone derivatives by nickel-catalyzed annulation of 2-halobenzamides with alkynes.
Organic letters (2010-07-09)
Chuan-Che Liu, Kanniyappan Parthasarathy, Chien-Hong Cheng
ABSTRACT
An efficient method for the synthesis of substituted 1(2H)-isoquinolone derivatives via nickel-catalyzed annulation of substituted 2-halobenzamides with alkynes is described. This protocol is successfully applied to the total synthesis of oxyavicine with excellent yield.
Sigma-Aldrich
[1,2-Bis(diphenylphosphino)ethane]dibromonickel(II), ≥95%
Pricing and availability is not currently available.