Merck
  • Regulatory action of prolactin on the in vitro growth of CD34+ve human hemopoietic progenitor cells.
Regulatory action of prolactin on the in vitro growth of CD34+ve human hemopoietic progenitor cells.
Journal of cellular physiology (1995-05-01)
G Bellone, M Geuna, A Carbone, S Silvestri, R Foa, G Emanuelli, L Matera
ABSTRACT
The pituitary hormone prolactin (Prl) is known to act as a local regulator of immune cell function, and Prl-binding receptors (Prl-R) have been described to share distinctive features with the members of the newly described cytokine/hemopoietin receptor superfamily. Here we show that the hormone can functionally interact with lineage-specific hemopoietic factors. When highly purified progenitor cells (CD34+ve) were seeded in semisolid methylcellulose cultures in the presence of interleukin (IL)-3, granulocyte-macrophage colony stimulating factor (GM-CSF), and erythropoietin (Epo), a selective enhancing effect of Prl on the formation of colony forming unit-granulocyte (CFU-G) and burst forming unit-erythroid (BFU-E) colonies was observed. The effect of the hormone was plotted as a bell shaped curve, with the optimal response at the supraphysiological concentration of 50 ng/ml. Limiting dilution analysis showed that Prl acted directly on hemopoietic progenitors. This was confirmed by the observation on the CD34+ve cells of Prl-binding sites reacting with the specific monoclonal antibodies (mAbs), U5 and PrR-7A. Immunoprecipitation of the metabolically labeled CD34+ve cells with the PrR-7A mAb revealed a structure of 43 kD under reducing conditions. Analysis of the early events associated with the Prl/Prl-R interaction showed an increased number of cells engaged in DNA and hemoglobin synthesis. Enhanced erythroid differentiation of CD34+ve cells in the presence of Prl was secondary to upmodulation of receptors for the lineage-specific factor Epo. Together these data demonstrate the existence of a functional interplay between Prl and hemopoietic factors.
Sigma-Aldrich
Prolactin human, recombinant, expressed in E. coli, lyophilized powder, BioReagent, suitable for cell culture, >97% (SDS-PAGE)
Pricing and availability is not currently available.