- Small-molecule activation of neuronal cell fate.
Small-molecule activation of neuronal cell fate.
Nature chemical biology (2008-06-17)
Jay W Schneider, Zhengliang Gao, Shijie Li, Midhat Farooqi, Tie-Shan Tang, Ilya Bezprozvanny, Doug E Frantz, Jenny Hsieh
PMID18552832
ABSTRACT
We probed an epigenetic regulatory path from small molecule to neuronal gene activation. Isoxazole small molecules triggered robust neuronal differentiation in adult neural stem cells, rapidly signaling to the neuronal genome via Ca(2+) influx. Ca(2+)-activated CaMK phosphorylated and mediated nuclear export of the MEF2 regulator HDAC5, thereby de-repressing neuronal genes. These results provide new tools to explore the epigenetic signaling circuitry specifying neuronal cell fate and new leads for neuro-regenerative drugs.