Merck
  • Binding preference of nitroimidazolic radiosensitizers to nucleobases and nucleosides probed by electrospray ionization mass spectrometry and density functional theory.
Binding preference of nitroimidazolic radiosensitizers to nucleobases and nucleosides probed by electrospray ionization mass spectrometry and density functional theory.
The Journal of chemical physics (2019-01-10)
S Pandeti, L Feketeová, T J Reddy, H Abdoul-Carime, B Farizon, M Farizon, T D Märk
ABSTRACT
Nitroimidazolic radiosensitizers are used in radiation therapy to selectively sensitize cancer cells deprived of oxygen, and the actual mechanism of radiosensitization is still not understood. Selecting five radiosensitizers (1-methyl-5-nitroimidazole, ronidazole, ornidazole, metronidazole, and nimorazole) with a common 5-nitroimidazolic ring with different substitutions at N1 and C2 positions of the imidazole moiety, we investigate here their binding to nucleobases (A, T, G, and C) and nucleosides (As, Td, Gs, and Cd) via the positive electrospray ionization mass spectrometry experiments. In addition, quantum chemical calculations at the M062x/6-311+G(d,p) level of theory and basis set were used to determine binding energies of the proton bound dimers of a radiosensitizer and a nucleobase. The positive electrospray ionization leads to the formation of proton bound dimers of all radiosensitizers except 1-methyl-5-nitroimidazole in high abundance with C and smaller abundance with G. Ronidazole and metronidazole formed less abundant dimers also with A, while no dimers were observed to be formed at all with T. In contrast to the case of the nucleoside Td, the dimer intensity is as high as that with Cd, while the abundance of the dimer with Gs is smaller than that of the former. The experimental results are consistent with the calculations of binding energies suggesting proton bound dimers with C and G to be the strongest bound ones. Finally, a barrier-free proton transfer is observed when protonated G or C approaches the nitroimidazole ring.