Merck
  • Analysis of phototoxin taste closely correlates nucleophilicity to type 1 phototoxicity.
Analysis of phototoxin taste closely correlates nucleophilicity to type 1 phototoxicity.
Proceedings of the National Academy of Sciences of the United States of America (2019-05-30)
Eun Jo Du, Tae Jung Ahn, Hwajin Sung, HyunJi Jo, Hyung-Wook Kim, Seong-Tae Kim, KyeongJin Kang
ABSTRACT
Pigments often inflict tissue-damaging and proaging toxicity on light illumination by generating free radicals and reactive oxygen species (ROS). However, the molecular mechanism by which organisms sense phototoxic pigments is unknown. Here, we discover that Transient Receptor Potential Ankyrin 1-A isoform [TRPA1(A)], previously shown to serve as a receptor for free radicals and ROS induced by photochemical reactions, enables Drosophila melanogaster to aphotically sense phototoxic pigments for feeding deterrence. Thus, TRPA1(A) detects both cause (phototoxins) and effect (free radicals and ROS) of photochemical reactions. A group of pigment molecules not only activates TRPA1(A) in darkness but also generates free radicals on light illumination. Such aphotic detection of phototoxins harboring the type 1 (radical-generating) photochemical potential requires the nucleophile-sensing ability of TRPA1. In addition, agTRPA1(A) from malaria-transmitting mosquitoes Anopheles gambiae heterologously produces larger current responses to phototoxins than Drosophila TRPA1(A), similar to their disparate nucleophile responsiveness. Along with TRPA1(A)-stimulating capabilities, type 1 phototoxins exhibit relatively strong photo-absorbance and low energy gaps between the highest occupied molecular orbital and the lowest unoccupied molecular orbital. However, TRPA1(A) activation is more highly concordant to type 1 phototoxicity than are those photochemical parameters. Collectively, nucleophile sensitivity of TRPA1(A) allows flies to taste potential phototoxins for feeding deterrence, preventing postingestive photo-injury. Conversely, pigments need to bear high nucleophilicity (electron-donating propensity) to act as type 1 phototoxins, which is consistent with the fact that transferring photoexcited electrons from phototoxins to other molecules causes free radicals. Thus, identification of a sensory mechanism in Drosophila reveals a property fundamental to type 1 phototoxins.