Merck
  • Temperature Responsive Nanoparticles Based on PEGylated Polyaspartamide Derivatives for Drug Delivery.
Temperature Responsive Nanoparticles Based on PEGylated Polyaspartamide Derivatives for Drug Delivery.
Polymers (2019-04-10)
Guangyan Zhang, Xulin Jiang
ABSTRACT
The temperature responsive PEGylated polyaspartamide derivative, denoted as mPEG-PAAHP, was synthesized by the click reaction. FTIR and ¹H NMR were adopted to characterize and confirm the chemical structures of the obtained mPEG-PAAHPs. The temperature responsive behavior investigated by transmittance and dynamic light scattering showed that some of the obtained mPEG-PAAHPs exhibited obvious temperature responsiveness and could be used to prepare nanoparticles by quickly heating. Drug paclitaxel can be encapsulated into mPEG-PAAHP based nanoparticles with a high encapsulation efficiency up to 99% (corresponding to a drug loading content of around 9.9%). Dynamic light scattering results showed that the PTX-loaded nanoparticles had a mean size around 80 nm (PDI<0.2) and good stability in PBS with 150 mM ionic strength. In vitro cytotoxicity results showed that mPEG-PAAHP did not show any toxicity to HeLa cells, but the PTX-loaded nanoparticles based on mPEG-PAAHP exhibited obvious anti-cancer activity. Thus, the temperature responsive PEGylated polyaspartamide derivative mPEG-PAAHP may be a promising drug delivery system.