- Effects of charge balance and hydrophobicity of the surface of cytochrome c on the distribution behaviour in an ionic liquid/buffer biphasic system.
Effects of charge balance and hydrophobicity of the surface of cytochrome c on the distribution behaviour in an ionic liquid/buffer biphasic system.
Factors contributing to the different distribution behaviour of cytochrome c were investigated in a biphasic tetrabutylphosphonium 2,4,6-trimethylbenzenesulfonate and potassium phosphate buffer system, which shows a lower critical solution temperature. To change charge balance and hydrophobicity of cytochrome c, surface modification with a few modifier molecules was applied. Surface charge and hydrophobicity affected the distribution behavior of chemically modified cytochrome c in the tetrabutylphosphonium 2,4,6-trimethylbenzenesulfonate and potassium phosphate buffer biphasic system. The distribution ratio into tetrabutylphosphonium 2,4,6-trimethylbenzenesulfonate decreased with decreasing isoelectric point of cytochrome c. Furthermore, cytochrome c possessing a low isoelectric point showed different distribution ratio depending on surface hydrophobicity. Taken together, these findings indicate that isoelectric point and surface hydrophobicity of cytochrome c are important factors controlling the distribution behavior in temperature sensitive biphasic systems.