- Tailoring Rheological Properties of Thermoresponsive Hydrogels through Block Copolymer Adsorption to Cellulose Nanocrystals.
Tailoring Rheological Properties of Thermoresponsive Hydrogels through Block Copolymer Adsorption to Cellulose Nanocrystals.
This study investigates the adsorption of a block copolymer composed of a poly(2-(dimethylamino)ethyl methacrylate) (PDMAEMA) cationic polyelectrolyte and a poly(di(ethylene glycol) methyl ethermethacrylate) (PDEGMA) on oxidized cellulose nanocrystals (TO-CNCs) to produce hydrogels. PDMEAMA- b-PDEGMA was synthesized by atom-transfer radical polymerization. The extent and dynamics of the adsorption of PDMAEMA- b-PDEGMA on TO-CNCs were determined by electromechanical microbalance and optical techniques. Electrostatic adsorption was identified on TO-CNCs with the quaternized block copolymer. Small-angle neutron scattering experiments were performed to investigate the polymer behavior on the TO-CNC surfaces. Depending on the temperature, block copolymer induces the aggregation of nanocrystals after adsorption by connecting CNCs bundles with block copolymer chains. A reversible liquid-to-gel transition, triggered by temperature, was clearly detected by rheological measurements for the copolymer-CNC mixtures. At the optimal copolymer to CNC ratio the viscosity increased by 4 orders of magnitude at low shear rates. These stimuli-responsive CNC-based materials could be used as injectable biomedical systems.