Merck
  • Investigations on the Maillard Reaction in Sesame ( Sesamum indicum L.) Seeds Induced by Roasting.
Investigations on the Maillard Reaction in Sesame ( Sesamum indicum L.) Seeds Induced by Roasting.
Journal of agricultural and food chemistry (2019-04-11)
Ecem Berk, Aytül Hamzalıoğlu, Vural Gökmen
ABSTRACT
This study investigated the formation of Maillard reaction products in sesame seeds under different roasting conditions. Sesame seeds were roasted at 150, 180, 200, and 220 °C for 10 min, and thermal process contaminants including 5-hydroxymethylfurfural, acrylamide, furan, and dicarbonyl compounds (1-deoxyglucosone, 3-deoxyglucosone, methylglyoxal, and diacetyl) together with glycation markers namely N-ε-fructosyllysine, N-ε-carboxymethyllysine, and N-ε-carboxyethyllysine, were monitored. Roasting induced the formation of 5-hydroxymethylfurfural, acrylamide, and dicarbonyl compounds, except furan, significantly ( p < 0.05). 5-Hydroxymethylfurfural and acrylamide content of roasted sesame seeds were found to vary as 3-40 mg/kg and 135-633 μg/kg, respectively. Dicarbonyl compounds were in the following order: methylglyoxal > 3-deoxyglucosone > 1-deoxyglucosone > diacetyl. On the other hand, N-ε-fructosyllysine concentration decreased while the roasting temperature increased; however, N-ε-carboxymethyllysine and N-ε-carboxyethyllysine formation was induced under those conditions. This is the first study reporting the formation of thermal process contaminants and glycation markers in sesame seeds through Maillard reaction during roasting.