Merck
Antiviral activity of the photoactive plant pigment hypericin.
Photochemistry and photobiology (1991-07-01)
I Lopez-Bazzocchi, J B Hudson, G H Towers
ABSTRACT
The polycyclic compound hypericin, a known photodynamic agent, was investigated for antiviral activity in the presence and absence of light. The three viruses tested: murine cytomegalovirus; Sindbis virus; and human immunodeficiency virus type 1, were all susceptible to hypericin; but these antiviral activities were considerably enhanced, in a dose-dependent manner, by exposure to light.
Sigma-Aldrich
Hypericin from Hypericum perforatum, ~95% (HPLC)
Pricing and availability is not currently available.