Merck
  • Effect of aliphatic diamine spacer length on enzymatic performance of myrosinase immobilized on chitosan microsphere and its application for sulforaphene production.
Effect of aliphatic diamine spacer length on enzymatic performance of myrosinase immobilized on chitosan microsphere and its application for sulforaphene production.
Journal of biotechnology (2019-05-03)
Jie Zhang, Chunrong Feng, Xuemei Tan, Peter-Leon Hagedoorn, Changhong Gu, Hui Xu, Xing Zhou
ABSTRACT
Glucosinolates can be hydrolyzed by the enzyme commonly known as myrosinase (E.C. 3.2.1.147) to a variety of biological compounds. Myrosinase (MYR) has been immobilized through the flexible spacers of different length on cross-linked chitosan resin (CCR). Ethylene diamine, hexamethylenediamine and decamethylene-diamine have been separately used as spacers. The influence of the flexible spacer length on the immobilized MYR (IMYR) properties were evaluated. The optimum pH and Vmax of IMYR linearly increase with the flexible spacer length, and the optimum temperature and Km of IMYR show an opposite trend. The recyclability of IMYR was good, with 90% recovery of activity after 10 cycles and 80% recovery after 30 cycles. IMYR was highly stable under storage conditions, with 95% recovery of activity after one year storage at 4 °C. The IMYR with the longest flexible spacer, decamethylene-diamine, was used as a biocatalyst for sulforaphene production. The overall hydrolysis ratio of glucoraphenin was 93.25 ± 0.91% and the activity of DDMCCR-IMYR remained 95% after 10 days of continuous use.