Merck
  • Silencing synaptic MicroRNA-411 reduces voluntary alcohol consumption in mice.
Silencing synaptic MicroRNA-411 reduces voluntary alcohol consumption in mice.
Addiction biology (2018-04-18)
Dana Most, Nihal A Salem, Gayatri R Tiwari, Yuri A Blednov, R Dayne Mayfield, R Adron Harris
ABSTRACT
Chronic alcohol consumption alters the levels of microRNAs and mRNAs in the brain, but the specific microRNAs and processes that target mRNAs to affect cellular function and behavior are not known. We examined the in vivo manipulation of previously identified alcohol-responsive microRNAs as potential targets to reduce alcohol consumption. Silencing of miR-411 by infusing antagomiR-411 into the prefrontal cortex of female C57BL/6J mice reduced alcohol consumption and preference, without altering total fluid consumption, saccharin consumption, or anxiety-related behaviors. AntagomiR-411 reduced alcohol consumption when given to mice exposed to a chronic alcohol drinking paradigm but did not affect the acquisition of consumption in mice without a history of alcohol exposure, suggesting that antagomiR-411 has a neuroadaptive, alcohol-dependent effect. AntagomiR-411 decreased the levels of miR-411, as well as the association of immunoprecipitated miR-411 with Argonaute2; and, it increased levels of Faah and Ppard mRNAs. Moreover, antagomiR-411 increased the neuronal expression of glutamate receptor AMPA-2 protein, a known alcohol target and a predicted target of miR-411. These results suggest that alcohol and miR-411 function in a homeostatic manner to regulate synaptic mRNA and protein, thus reversing alcohol-related neuroadaptations and reducing chronic alcohol consumption.
Sigma-Aldrich
Monoclonal Anti-AGO2 antibody produced in rat, ~1.5 mg/mL, clone 11A9, purified immunoglobulin
Pricing and availability is not currently available.
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Pricing and availability is not currently available.