Merck
  • Two Distinct E2F Transcriptional Modules Drive Cell Cycles and Differentiation.
Two Distinct E2F Transcriptional Modules Drive Cell Cycles and Differentiation.
Cell reports (2019-05-28)
Maria C Cuitiño, Thierry Pécot, Daokun Sun, Raleigh Kladney, Takayuki Okano-Uchida, Neelam Shinde, Resham Saeed, Antonio J Perez-Castro, Amy Webb, Tom Liu, Soo In Bae, Linda Clijsters, Nicholas Selner, Vincenzo Coppola, Cynthia Timmers, Michael C Ostrowski, Michele Pagano, Gustavo Leone
ABSTRACT
Orchestrating cell-cycle-dependent mRNA oscillations is critical to cell proliferation in multicellular organisms. Even though our understanding of cell-cycle-regulated transcription has improved significantly over the last three decades, the mechanisms remain untested in vivo. Unbiased transcriptomic profiling of G0, G1-S, and S-G2-M sorted cells from FUCCI mouse embryos suggested a central role for E2Fs in the control of cell-cycle-dependent gene expression. The analysis of gene expression and E2F-tagged knockin mice with tissue imaging and deep-learning tools suggested that post-transcriptional mechanisms universally coordinate the nuclear accumulation of E2F activators (E2F3A) and canonical (E2F4) and atypical (E2F8) repressors during the cell cycle in vivo. In summary, we mapped the spatiotemporal expression of sentinel E2F activators and canonical and atypical repressors at the single-cell level in vivo and propose that two distinct E2F modules relay the control of gene expression in cells actively cycling (E2F3A-8-4) and exiting the cycle (E2F3A-4) during mammalian development.
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, Mitosis Marker, Upstate®, from rabbit
Sigma-Aldrich
Anti-E2F4 Antibody, clone LLF4-2, clone LLF4-2, from mouse
Sigma-Aldrich
Protease Inhibitor Cocktail, for use in tissue culture media, DMSO solution