Merck
  • Ruthenium-NHC-Diamine Catalyzed Enantioselective Hydrogenation of Isocoumarins.
Ruthenium-NHC-Diamine Catalyzed Enantioselective Hydrogenation of Isocoumarins.
Journal of the American Chemical Society (2017-02-02)
Wei Li, Mario P Wiesenfeldt, Frank Glorius
ABSTRACT
A novel and practical chiral ruthenium-NHC-diamine system is disclosed for the enantioselective hydrogenation of isocoumarins, which provides a new concept to apply (chiral) NHC ligands in asymmetric catalysis. A variety of optically active 3-substituted 3,4-dihydroisocoumarins were obtained in excellent enantioselectivities (up to 99% ee). Moreover, this methodology was utilized in the synthesis of O-methylmellein, mellein, and ochratoxin A.
Sigma-Aldrich
(R,R)-SINpEt⋅HBF4, ≥95%