Merck
  • Effects of amelogenins on angiogenesis-associated processes of endothelial cells.
Effects of amelogenins on angiogenesis-associated processes of endothelial cells.
Journal of wound care (2011-03-08)
S Almqvist, H K Kleinman, M Werthén, P Thomsen, M S Agren
ABSTRACT
To study the effects of an amelogenin mixture on integrin-dependent adhesion, DNA synthesis and apoptosis of cultured human dermal microvascular endothelial cells and angiogenesis in an organotypic assay. Immobilised antibodies against specific integrins (alpha-1, alpha-2, alpha-3, alpha-4, alpha-5, alpha-v, ß1, ß2, ß3, ß4, ß6, alpha-vß3, alpha-vß5 and alpha-5ß1) were used to capture treated human dermal microvascular endothelial cells, which were detected colourimetrically. DNA synthesis of the cells was monitored by 5-bromo-2'- deoxyuridine incorporation and apoptosis by a TdT-mediated dUTP nick-end labelling technique. Tubule formation from aortic arches of 13-d-old chick embryos were followed over 48h. The amelogenin mixture increased microvessel outgrowth by 76% (p < 0.01, n=12) from the aortic explants. Also, amelogenins increased the adhesion (p < 0.01, n = 5) by multiple angiogenesis associated integrin subunits and alpha-vß3, alpha-vß5 and alpha-5ß1 heterodimers on human dermal microvascular endothelial cells at a non-mitogenic concentration (100 µg/ml). Conversely, amelogenins at 1,000 µg/ml decreased microvessel formation possibly due to attenuation of corresponding integrins despite increasing (p < 0.001, n = 8) DNA synthesis. No significant apoptosis was detected in human dermal microvascular endothelial cells cultured on Matrigel with and without amelogenins. Increased surface expression of integrins on endothelial cells may contribute to the proangiogenic property of amelogenins.
Sigma-Aldrich
α/β Integrin-mediated Cell Adhesion Array Combo Kit, colorimetic, The Alpha/Beta Integrin-Mediated Cell Adhesion Array Kit can be used for assessing the presence or absence of specific integrins on the cell surface.
Pricing and availability is not currently available.