Merck
  • Purification and characterization of dipeptidyl peptidase I from human spleen.
Purification and characterization of dipeptidyl peptidase I from human spleen.
Archives of biochemistry and biophysics (1992-06-01)
M J McGuire, P E Lipsky, D L Thiele
ABSTRACT
The lysosomal hydrolase, dipeptidyl peptidase I (DPPI), was purified from human spleen and its enzymatic activity characterized. The enzyme was purified to apparent homogeneity by a combination of differential pH solubility, heat-treatment, affinity chromatography on concanavalin A-agarose and p-hydroxymercuribenzoate-agarose, and gel filtration chromatography on Sephacryl S-300. This procedure resulted in a 1100-fold purification of DPPI protein with a yield of approximately 2% of the total DPPI activity. The enzyme was characterized as a glycoprotein with a pI of 5.4, a molecular mass of 200,000 Da as determined by gel filtration under nondenaturing conditions, and a subunit size of 24,000 Da. Amino acid sequence analysis of peptides isolated from cyanogen bromide and trypsin digests of the 24,000-Da subunit revealed extensive sequence similarity between human and rat DPPI. Purified DPPI exhibited both hydrolytic and transpeptidase (polymerase) activity. DPPI exhibited activity against a variety of dipeptide substrates including peptides with either non-polar or polar residues in the P1 position. In contrast to the reported substrate specificity of bovine and murine DPPI, the human enzyme exhibited a modest preference for peptides with nonpolar residues in the P1 position. DPPI content was found to be highest among cytotoxic lymphocytes and myeloid cells. The high level of DPPI expression in these cell populations correlates with their sensitivity to the toxic effects of leucyl-leucine methyl ester, a substrate for DPPI.
Sigma-Aldrich
Gly-Arg 4-methoxy-β-naphthylamide dihydrochloride, dipeptidyl aminopeptidase I substrate
Pricing and availability is not currently available.
Sigma-Aldrich
Ser-Tyr β-naphthylamide, powder
Pricing and availability is not currently available.