Merck
  • The AMPA receptor GluR2 C terminus can mediate a reversible, ATP-dependent interaction with NSF and alpha- and beta-SNAPs.
The AMPA receptor GluR2 C terminus can mediate a reversible, ATP-dependent interaction with NSF and alpha- and beta-SNAPs.
Neuron (1998-08-11)
P Osten, S Srivastava, G J Inman, F S Vilim, L Khatri, L M Lee, B A States, S Einheber, T A Milner, P I Hanson, E B Ziff
ABSTRACT
In this study, we demonstrate specific interaction of the GluR2 alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionate (AMPA) receptor subunit C-terminal peptide with an ATPase N-ethylmaleimide-sensitive fusion protein (NSF) and alpha- and beta-soluble NSF attachment proteins (SNAPs), as well as dendritic colocalization of these proteins. The assembly of the GluR2-NSF-SNAP complex is ATP hydrolysis reversible and resembles the binding of NSF and SNAP with the SNAP receptor (SNARE) membrane fusion apparatus. We provide evidence that the molar ratio of NSF to SNAP in the GluR2-NSF-SNAP complex is similar to that of the t-SNARE syntaxin-NSF-SNAP complex. NSF is known to disassemble the SNARE protein complex in a chaperone-like interaction driven by ATP hydrolysis. We propose a model in which NSF functions as a chaperone in the molecular processing of the AMPA receptor.
Sigma-Aldrich
Anti-NMDAR2A Antibody, serum, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Syntaxin Antibody, clone SP8, ascites fluid, clone SP8, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Heat Shock Protein 70 antibody produced in mouse, clone BRM-22, ascites fluid
Pricing and availability is not currently available.