Merck
  • Aire deficient mice develop multiple features of APECED phenotype and show altered immune response.
Aire deficient mice develop multiple features of APECED phenotype and show altered immune response.
Human molecular genetics (2002-02-21)
Chris Ramsey, Ola Winqvist, Lea Puhakka, Maria Halonen, Aune Moro, Olle Kämpe, Petra Eskelin, Markku Pelto-Huikko, Leena Peltonen
ABSTRACT
Autoimmune polyendocrinopathy-candidiasis-ectodermal dystrophy (APECED) is a monogenic autosomal recessive disease caused by mutations in the AIRE gene. Here we have produced knock-out mice for the Aire gene. The Aire-/- mice develop normally; however, autoimmune features of APECED in Aire-/- mice are evident, including multiorgan lymphocytic infiltration, circulating autoantibodies and infertility. The distribution of B and T cells and thymic maturation as well as activation of T cells appear normal, while the TCR-Vbeta repertoire is altered in peripheral T cells of Aire-/- mice. When mice are challenged with immunization, the peripheral T cells of Aire-/- mice have a 3-5-fold increased proliferation. These findings suggest that the Aire gene is not necessary for normal T cell education and development, while a defect in immune response detected in challenged Aire-/- mice underlines the crucial role of AIRE/Aire in maintaining homeostatic regulation in the immune system.