Merck
  • scAAV-mediated gene transfer of interleukin-1-receptor antagonist to synovium and articular cartilage in large mammalian joints.
scAAV-mediated gene transfer of interleukin-1-receptor antagonist to synovium and articular cartilage in large mammalian joints.
Gene therapy (2012-11-16)
R S Watson, T A Broome, P P Levings, B L Rice, J D Kay, A D Smith, E Gouze, J-N Gouze, E A Dacanay, W W Hauswirth, D M Nickerson, M J Dark, P T Colahan, S C Ghivizzani
ABSTRACT
With the long-term goal of developing a gene-based treatment for osteoarthritis (OA), we performed studies to evaluate the equine joint as a model for adeno-associated virus (AAV)-mediated gene transfer to large, weight-bearing human joints. A self-complementary AAV2 vector containing the coding regions for human interleukin-1-receptor antagonist (hIL-1Ra) or green fluorescent protein was packaged in AAV capsid serotypes 1, 2, 5, 8 and 9. Following infection of human and equine synovial fibroblasts in culture, we found that both were only receptive to transduction with AAV1, 2 and 5. For these serotypes, however, transgene expression from the equine cells was consistently at least 10-fold higher. Analyses of AAV surface receptor molecules and intracellular trafficking of vector genomes implicate enhanced viral uptake by the equine cells. Following delivery of 1 × 10(11) vector genomes of serotypes 2, 5 and 8 into the forelimb joints of the horse, all three enabled hIL-1Ra expression at biologically relevant levels and effectively transduced the same cell types, primarily synovial fibroblasts and, to a lesser degree, chondrocytes in articular cartilage. These results provide optimism that AAV vectors can be effectively adapted for gene delivery to large human joints affected by OA.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, Phycoerythrin conjugated, clone LM609, Chemicon®, from mouse