Merck
Protein measurement with the Folin phenol reagent.
The Journal of biological chemistry (1951-11-01)
O H LOWRY, N J ROSEBROUGH, A L FARR, R J RANDALL
Sigma-Aldrich
Total Protein Kit, Micro Lowry, Peterson′s Modification
Sigma-Aldrich
Bicinchoninic Acid Kit for Protein Determination, for 200-1000 μg/ml protein
Sigma-Aldrich
Folin & Ciocalteu′s phenol reagent, suitable for determination of total protein by Lowry method, 1.9-2.1 N