Merck
  • Sarcosine in prostate cancer tissue is not a differential metabolite for prostate cancer aggressiveness and biochemical progression.
Sarcosine in prostate cancer tissue is not a differential metabolite for prostate cancer aggressiveness and biochemical progression.
The Journal of urology (2010-12-21)
Florian Jentzmik, Carsten Stephan, Michael Lein, Kurt Miller, Beate Kamlage, Bianca Bethan, Glen Kristiansen, Klaus Jung
ABSTRACT
Sarcosine in prostate cancer tissue samples was recently reported to be increased during prostate cancer progression to metastasis and suggested to be a key metabolite of cancer cell invasion and aggressiveness. We reevaluated sarcosine in prostate cancer tissue samples as a potential indicator of tumor aggressiveness, and as a predictor of recurrence-free survival. Sarcosine in matched samples of malignant and nonmalignant tissue from 92 patients with prostate cancer after radical prostatectomy was measured in the framework of a global metabolite profiling study of prostate cancer by gas chromatography/mass spectrometry. We related results to age, prostate volume, tumor stage, Gleason score, preoperative prostate specific antigen and biochemical recurrence, defined as a persistent prostate specific antigen increase of greater than 0.2 ng/ml. Nonparametric statistical tests, ROC curves and Kaplan-Meier analyses were done. Median sarcosine content in tissue was about 7% higher in matched malignant vs nonmalignant samples, which was significantly. Sarcosine values were not associated with tumor stage (pT2 vs pT3), tumor grade (Gleason score less than 7 vs 7 or greater) or biochemical recurrence. The lack of metastatic tissue samples was a study limitation. Sarcosine in prostate cancer tissue samples cannot be considered a suitable predictor of tumor aggressiveness or biochemical recurrence.
Sigma-Aldrich
Sarcosine, BioXtra
Pricing and availability is not currently available.