Merck
  • Enhancement of colloid uptake by tumor cell surface electrical charge modification.
Enhancement of colloid uptake by tumor cell surface electrical charge modification.
Biochimica et biophysica acta (1989-07-10)
L J Anghileri, P Maincent, J Robert
ABSTRACT
Colloidal [51Cr]chromic phosphate uptake is considerably increased by preincubation of P388 ascites leukemia cells with poly(DL-lysine). The uptake increase is in direct relationship with the concentration and the degree of polymerization of poly(DL-lysine). The probable implication of cell surface electrical charge modification in these phenomena is discussed.
Sigma-Aldrich
Poly-DL-lysine hydrobromide, mol wt >40,000
Pricing and availability is not currently available.
Sigma-Aldrich
Poly-DL-lysine hydrobromide, mol wt 25,000-40,000
Pricing and availability is not currently available.