Merck
  • Purification and characterization of the acetyl-CoA synthetase from Mycobacterium tuberculosis.
Purification and characterization of the acetyl-CoA synthetase from Mycobacterium tuberculosis.
Acta biochimica et biophysica Sinica (2011-09-08)
Ru Li, Jing Gu, Peng Chen, Zhiping Zhang, Jiaoyu Deng, Xianen Zhang
ABSTRACT
Acetyl-CoA (AcCoA) synthetase (Acs) catalyzes the conversion of acetate into AcCoA, which is involved in many catabolic and anabolic pathways. Although this enzyme has been studied for many years in many organisms, the properties of Mycobacterium tuberculosis Acs and the regulation of its activity remain unknown. Here, the putative acs gene of M. tuberculosis H37Rv (Mt-Acs) was expressed as a fusion protein with 6×His-tag on the C-terminus in Escherichia coli. The recombinant Mt-Acs protein was successfully purified and then its enzymatic characteristics were analyzed. The optimal pH and temperature, and the kinetic parameters of Mt-Acs were determined. To investigate whether Mt-Acs is regulated by lysine acetylation as reported for Salmonella enterica Acs, its mutant K617R was also generated. Determination of the enzymatic activity suggests that Lys-617 is critical for its function. We further demonstrated that Mt-Acs underwent auto-acetylation with acetate but not with AcCoA as the acetyl donor, which resulted in the decrease of its activity. CoA, the substrate for AcCoA formation, inhibited the auto-acetylation. Furthermore, the silent information regulator (Sir2) of M. tuberculosis (Mt-Sir2) could catalyze Mt-Acs deacetylation, which resulted in activation of Acs. These results may provide more insights into the physiological roles of Mt-Acs in M. tuberculosis central metabolism.
Sigma-Aldrich
Acetyl coenzyme A lithium salt, ≥93% (HPLC)
Sigma-Aldrich
Acetyl coenzyme A trisodium salt, ≥93% (HPLC), powder
Sigma-Aldrich
S-Acetyl-coenzyme A synthetase from baker′s yeast (S. cerevisiae), lyophilized powder, ≥3 units/mg protein