Merck
  • Cytoplasmic SET induces tau hyperphosphorylation through a decrease of methylated phosphatase 2A.
Cytoplasmic SET induces tau hyperphosphorylation through a decrease of methylated phosphatase 2A.
BMC neuroscience (2014-07-02)
Stéphanie Chasseigneaux, Christine Clamagirand, Léa Huguet, Lucie Gorisse-Hussonnois, Christiane Rose, Bernadette Allinquant
ABSTRACT
The neuronal cytoplasmic localization of SET, an inhibitor of the phosphatase 2A (PP2A), results in tau hyperphosphorylation in the brains of Alzheimer patients through mechanisms that are still not well defined. We used primary neurons and mouse brain slices to show that SET is translocated to the cytoplasm in a manner independent of both its cleavage and over-expression. The localization of SET in the cytoplasm, either by the translocation of endogenous SET or by internalization of the recombinant full-length SET protein, induced tau hyperphosphorylation. Cytoplasmic recombinant full-length SET in mouse brain slices induced a decrease of PP2A activity through a decrease of methylated PP2A levels. The levels of methylated PP2A were negatively correlated with tau hyperphosphorylation at Ser-202 but not with the abnormal phosphorylation of tau at Ser-422. The presence of full-length SET in the neuronal cytoplasm is sufficient to impair PP2A methylation and activity, leading to tau hyperphosphorylation. In addition, our data suggest that tau hyperphosphorylation is regulated by different mechanisms at distinct sites. The translocation of SET to the neuronal cytoplasm, the low activity of PP2A, and tau hyperphosphorylation are associated in the brains of Alzheimer patients. Our data show a link between the translocation of SET in the cytoplasm and the decrease of methylated PP2A levels leading to a decrease of PP2A activity and tau hyperphosphorylation. This chain of events may contribute to the pathogenesis of Alzheimer disease.
Sigma-Aldrich
Anti-PP2A Antibody, C subunit, clone 1D6, clone 1D6, Upstate®, from mouse
Sigma-Aldrich
Anti-methyl-PP2A Antibody, C subunit, clone 2A10, clone 2A10, from mouse
Sigma-Aldrich
Anti-Tau Mouse mAb (TAU-5), liquid, clone TAU-5, Calbiochem®