Merck
  • Testosterone improves the regeneration of old and young mouse skeletal muscle.
Testosterone improves the regeneration of old and young mouse skeletal muscle.
The journals of gerontology. Series A, Biological sciences and medical sciences (2012-04-14)
Carlo Serra, Frances Tangherlini, Sara Rudy, Daniel Lee, Gianluca Toraldo, Nicolae Lucian Sandor, Anqi Zhang, Ravi Jasuja, Shalender Bhasin
ABSTRACT
Aging is associated with loss of muscle mass and strength, reduced satellite cell number, and lower regenerative potential. Testosterone increases muscle mass, strength, and satellite cell number in humans; however, the effects of testosterone on the regenerative potential of skeletal muscle are unclear. Here, we investigated the effect of testosterone on the skeletal muscle regeneration of young (2-month-old) and aged (24-month-old) male mice. We show that testosterone increases the number of proliferating satellite cells in regenerating "tibialis anterior" muscle of young and aged castrated mice 2 and 4 days postinjury. Testosterone supplementation increases the number and the cross-sectional area of regenerating fibers in both classes of age 4 days postinjury. Testosterone increases satellite cell activation and proliferation and the regeneration of both young and aged mouse muscle. These data suggest prospective application of androgens to improve the regenerating potential of the aged human skeletal muscle.
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule Antibody, Chemicon®, from rabbit