Merck
  • Epigenetic repression of the Igk locus by STAT5-mediated recruitment of the histone methyltransferase Ezh2.
Epigenetic repression of the Igk locus by STAT5-mediated recruitment of the histone methyltransferase Ezh2.
Nature immunology (2011-11-01)
Malay Mandal, Sarah E Powers, Mark Maienschein-Cline, Elizabeth T Bartom, Keith M Hamel, Barbara L Kee, Aaron R Dinner, Marcus R Clark
ABSTRACT
During B lymphopoiesis, recombination of the locus encoding the immunoglobulin κ-chain complex (Igk) requires expression of the precursor to the B cell antigen receptor (pre-BCR) and escape from signaling via the interleukin 7 receptor (IL-7R). By activating the transcription factor STAT5, IL-7R signaling maintains proliferation and represses Igk germline transcription by unknown mechanisms. We demonstrate that a STAT5 tetramer bound the Igk intronic enhancer (E(κi)), which led to recruitment of the histone methyltransferase Ezh2. Ezh2 marked trimethylation of histone H3 at Lys27 (H3K27me3) throughout the κ-chain joining region (J(κ)) to the κ-chain constant region (C(κ)). In the absence of Ezh2, IL-7 failed to repress Igk germline transcription. H3K27me3 modifications were lost after termination of IL-7R-STAT5 signaling, and the transcription factor E2A bound E(κi), which resulted in acquisition of H3K4me1 and acetylated histone H4 (H4Ac). Genome-wide analyses showed a STAT5 tetrameric binding motif associated with transcriptional repression. Our data demonstrate how IL-7R signaling represses Igk germline transcription and provide a general model for STAT5-mediated epigenetic transcriptional repression.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit
Sigma-Aldrich
ChIPAb+ EED - ChIP Validated Antibody and Primer Set, from rabbit
Sigma-Aldrich
Anti-monomethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
ChIPAb+ SUZ12 - ChIP Validated Antibody and Primer Set, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®