Merck
  • Formation and maturation of parallel fiber-Purkinje cell synapses in the Staggerer cerebellum ex vivo.
Formation and maturation of parallel fiber-Purkinje cell synapses in the Staggerer cerebellum ex vivo.
The Journal of comparative neurology (2008-11-26)
Sonja Janmaat, Florence Frédéric, Klaas Sjollema, Paul Luiten, Jean Mariani, Johannes van der Want
ABSTRACT
In vivo, homozygous staggerer (Rora(sg/sg)) Purkinje cells (PCs) remain in an early stage of development with rudimentary spineless dendrites, associated with a lack of parallel fiber (PF) input and the persistence of multiple climbing fibers (CFs). In this immunocytochemical study we used cerebellar organotypic cultures to monitor the development of Rora(sg/sg) PF-PC synapses in the absence of CF innervation. Ex vivo the vesicular glutamate transporters VGluT1 and VGluT2 reactivity was preferentially localized around the Rora(sg/sg) PC soma and proximal dendrites, which are typically CF domains. The shift from VGluT2 to VGluT1 in PF terminals during development was delayed in Rora(sg/sg) slices. The postsynaptic receptors mGluR1 and GluRdelta2 were differently distributed on Rora(sg/sg) PCs. mGluR1 reactivity was evenly distributed in PC soma and dendrites, whereas GluRdelta2 reactivity, normally restricted at PF synapses, was dense in Rora(sg/sg) PC somata. The presynaptic distribution of VGluT1 and VGluT2 on Rora(sg/sg) PCs matched the postsynaptic distribution of the glutamate receptor GluRdelta2, but not mGluR1.
Sigma-Aldrich
Anti-Glutamate Receptor 1A, Metabotropic (mGluR1A) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.