Merck
  • Developmental maturation of excitation and inhibition balance in principal neurons across four layers of somatosensory cortex.
Developmental maturation of excitation and inhibition balance in principal neurons across four layers of somatosensory cortex.
Neuroscience (2010-12-01)
Z Zhang, Y-Y Jiao, Q-Q Sun
ABSTRACT
In adult cortices, the ratio of excitatory and inhibitory conductances (E/I ratio) is presumably balanced across a wide range of stimulus conditions. However, it is unknown how the E/I ratio is postnatally regulated, when the strength of synapses are rapidly changing. Yet, understanding of such a process is critically important, because there are numerous neuropsychological disorders, such as autism, epilepsy and schizophrenia, associated with disturbed E/I balances. Here we directly measured the E/I ratio underlying locally induced synaptic conductances in principal neurons from postnatal day 8 (P8) through 60. We found that (1) within each developmental period, the E/I ratio across four major cortical layers was maintained at a similar value under wide range of stimulation intensities; and (2) there was a rapid developmental decrease in the E/I ratio, which occurred within a sensitive period between P8 to P18 with exception of layer II/III. By comparing the excitatory and inhibitory conductances, as well as key synaptic protein expressions, we found a net increase in the number and strength of inhibitory, but not excitatory synapses, is responsible for the developmental decrease in the E/I ratio in the barrel cortex. The inhibitory markers were intrinsically co-regulated, gave rise to a sharp increase in the inhibitory conductance from P8 to P18. These results suggest that the tightly regulated E/I ratios in adults cortex is a result of drastic changes in relative weight of inhibitory but not excitatory synapses during critical period, and the local inhibitory structural changes are the underpinning of altered E/I ratio across postnatal development.
Sigma-Aldrich
Anti-Vesicular GABA Transporter (VGAT) Antibody, Chemicon®, from rabbit