Merck
  • Herpesvirus papio encodes a functional homologue of the Epstein-Barr virus apoptosis suppressor, BHRF1.
Herpesvirus papio encodes a functional homologue of the Epstein-Barr virus apoptosis suppressor, BHRF1.
The Journal of general virology (2000-06-22)
C A Meseda, J R Arrand, M Mackett
ABSTRACT
The human tumour virus Epstein-Barr virus (EBV) encodes a 17 kDa protein, BHRF1, which is a member of the BCL:-2 family and has been shown to suppress apoptosis. The role of this gene in the life-cycle of EBV has not been fully elucidated. In order to identify motifs conserved in herpesviruses and possibly shed light on its function we isolated a BHRF1 homologue from herpesvirus papio (cercopithecine herpesvirus-12) a closely related gammaherpesvirus of baboons. The gene, hvpBHRF1, also encodes a 17 kDa protein which shares 64% identity and 79% similarity with EBV BHRF1 at the amino acid level. In biological assays, hvpBHRF1 and BHRF1 conferred similar levels of protection on human keratinocytes induced to apoptose with cis-platin.
Sigma-Aldrich
Anti-EBV EA-R-p17 Antibody, clone 5B11, clone 5B11, Chemicon®, from mouse