Merck
  • Discovery of potent, selective, and orally bioavailable quinoline-based dipeptidyl peptidase IV inhibitors targeting Lys554.
Discovery of potent, selective, and orally bioavailable quinoline-based dipeptidyl peptidase IV inhibitors targeting Lys554.
Bioorganic & medicinal chemistry (2011-07-12)
Hironobu Maezaki, Yoshihiro Banno, Yasufumi Miyamoto, Yusuke Moritoh, Yuusuke Moritou, Tomoko Asakawa, Osamu Kataoka, Koji Takeuchi, Nobuhiro Suzuki, Koji Ikedo, Takuo Kosaka, Masako Sasaki, Shigetoshi Tsubotani, Akiyoshi Tani, Miyuki Funami, Yoshio Yamamoto, Michiko Tawada, Kathleen Aertgeerts, Jason Yano, Satoru Oi
ABSTRACT
Dipeptidyl peptidase IV (DPP-4) inhibition is a validated therapeutic option for type 2 diabetes, exhibiting multiple antidiabetic effects with little or no risk of hypoglycemia. In our studies involving non-covalent DPP-4 inhibitors, a novel series of quinoline-based inhibitors were designed based on the co-crystal structure of isoquinolone 2 in complex with DPP-4 to target the side chain of Lys554. Synthesis and evaluation of designed compounds revealed 1-[3-(aminomethyl)-4-(4-methylphenyl)-2-(2-methylpropyl)quinolin-6-yl]piperazine-2,5-dione (1) as a potent, selective, and orally active DPP-4 inhibitor (IC₅₀=1.3 nM) with long-lasting ex vivo activity in dogs and excellent antihyperglycemic effects in rats. A docking study of compound 1 revealed a hydrogen-bonding interaction with the side chain of Lys554, suggesting this residue as a potential target site useful for enhancing DPP-4 inhibition.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.995% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydroxylamine hydrochloride, 99.999% trace metals basis
Pricing and availability is not currently available.