Merck
  • Inhibition of lncRNA NEAT1 suppresses the inflammatory response in IBD by modulating the intestinal epithelial barrier and by exosome-mediated polarization of macrophages.
Inhibition of lncRNA NEAT1 suppresses the inflammatory response in IBD by modulating the intestinal epithelial barrier and by exosome-mediated polarization of macrophages.
International journal of molecular medicine (2018-08-23)
Rui Liu, Anliu Tang, Xiaoyan Wang, Xiong Chen, Lian Zhao, Zhiming Xiao, Shourong Shen
ABSTRACT
Inflammatory bowel disease (IBD) is a multifactorial inflammatory disease, and increasing evidence has demonstrated that the mechanism of the pathogenesis of IBD is associated with intestinal epithelial barrier injury. Long non‑coding RNAs (lncRNAs) are a class of transcripts >200 nucleotides in length with limited protein‑coding capability. Nuclear paraspeckle assembly transcript 1 (NEAT1) is a recently identified nuclear‑restricted lncRNA, which localizes in subnuclear structures, termed paraspeckles, and is involved in the immune response in a variety of ways. However, the function of NEAT1 in IBD remains to be fully elucidated. In the present study, reverse transcription‑quantitative polymerase chain reaction assays were performed to determine the expression levels of NEAT1 lncRNA in IBD serum samples and tissues. Furthermore, the effect of NEAT1 on the cell permeability of colon cells was investigated via determination of trans‑epithelial electrical resistance as well as performance of western blot and immunofluorescence assays. In addition, dextran sodium sulfate assays were performed to investigate the effect of downregulation of NEAT1 in IBD of mice. The present study detected the expression levels of NEAT1 in IBD cells and animal models to examine the changes in intestinal epithelial cell permeability following inhibition of the expression of NEAT1. In addition, phenotypic transformation was examined following different treatments in epithelial cells and macrophages. The results suggested that the expression of NEAT1 was high in IBD and was involved in the inflammatory response by regulating the intestinal epithelial barrier and through exosome‑mediated polarization of macrophages. The downregulation of NEAT1 suppressed the inflammatory response by modulating the intestinal epithelial barrier and through exosome‑mediated polarization of macrophages in IBD. The results of the present study revealed a potential strategy of targeting NEAT1 for IBD therapy.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
FITC-CM-Dextran, average mol wt 40,000
Pricing and availability is not currently available.