Merck
  • Application of Centrifugal Partition Chromatography for Bioactivity-Guided Purification of Antioxidant-Response-Element-Inducing Constituents from Atractylodis Rhizoma Alba.
Application of Centrifugal Partition Chromatography for Bioactivity-Guided Purification of Antioxidant-Response-Element-Inducing Constituents from Atractylodis Rhizoma Alba.
Molecules (Basel, Switzerland) (2018-09-12)
Myeong Il Kim, Ji Hoon Kim, Ahmed Shah Syed, Young-Mi Kim, Kevin Kyungsik Choe, Chul Young Kim
ABSTRACT
Activity-guided separation of antioxidant response element (ARE)-inducing constituents from the rhizomes of Atractylodis Rhizoma Alba was performed by the combination of centrifugal partition chromatography (CPC) and an ARE luciferase reporter assay. From 3 g of the active n-hexane fraction, one polyacetylene, (6E,12E)-tetradeca-6,12-dien-8,10-diyne-1,3-diyl diacetate (47.3 mg), and two sesquiterpenes, atractylenolide I (40.9 mg), and selina-4(14),7(11)-dien-8-one (6.0 mg) were successfully isolated by CPC with n-hexane⁻ethyl acetate⁻methanol⁻water (8:2:8:2, v/v). The chemical structures of the isolated compounds were determined by ¹H- and 13C-NMR and ESI-MS. Among the isolated compounds, (6E,12E)-tetradeca-6,12-diene-8,10-diyne-1,3-diol diacetate and selina-4(14),7(11)-dien-8-one increased ARE activity 32.9-fold and 16.6-fold, respectively, without significant cytotoxicity, when 5 µM sulforaphane enhanced ARE activity 27.1-fold. However, atractylenolide I did not increase ARE activity at 100 µM, and showed cytotoxicity at concentrations over 10 µM.
Sigma-Aldrich
DL-Sulforaphane, ≥90% (HPLC), synthetic, liquid