Merck
  • Crius: A novel fragment-based algorithm of de novo substrate prediction for enzymes.
Crius: A novel fragment-based algorithm of de novo substrate prediction for enzymes.
Protein science : a publication of the Protein Society (2018-05-04)
Zhiqiang Yao, Shuiqin Jiang, Lujia Zhang, Bei Gao, Xiao He, John Z H Zhang, Dongzhi Wei
ABSTRACT
The study of enzyme substrate specificity is vital for developing potential applications of enzymes. However, the routine experimental procedures require lot of resources in the discovery of novel substrates. This article reports an in silico structure-based algorithm called Crius, which predicts substrates for enzyme. The results of this fragment-based algorithm show good agreements between the simulated and experimental substrate specificities, using a lipase from Candida antarctica (CALB), a nitrilase from Cyanobacterium syechocystis sp. PCC6803 (Nit6803), and an aldo-keto reductase from Gluconobacter oxydans (Gox0644). This opens new prospects of developing computer algorithms that can effectively predict substrates for an enzyme.
Sigma-Aldrich
3-Methoxybutyl acetate, AldrichCPR
Pricing and availability is not currently available.
Sigma-Aldrich
3-Phenylpropyl acetate, ≥98%, FCC, FG
Pricing and availability is not currently available.
Sigma-Aldrich
2-Phenethyl acetate, 99%
Pricing and availability is not currently available.