Merck
  • The homeostasis-maintaining metabolites from bacterial stress response to bacteriophage infection suppress tumor metastasis.
The homeostasis-maintaining metabolites from bacterial stress response to bacteriophage infection suppress tumor metastasis.
Oncogene (2018-06-22)
Tianliang He, Min Jin, Chenxi Xu, Zhongjun Ma, Fufang Wu, Xiaobo Zhang
ABSTRACT
The antiviral metabolites from bacterial stress response to bacteriophage infection can maintain homeostasis of host cells, while metabolism disorder is a remarkable characteristic of tumorigenesis. In the aspect of metabolic homeostasis, therefore, the antiviral homeostasis-maintaining metabolites of bacteria may possess anti-tumor activity. However, this issue has not been addressed. Here we show that the homeostasis-challenged maintaining metabolites from deep-sea bacteriophage-challenged thermophile can suppress tumor metastasis. The results indicated that the metabolic profiles of the bacteriophage GVE2-infected and virus-free thermophile Geobacillus sp. E263 from a deep-sea hydrothermal vent were remarkably different. Thirteen metabolites were significantly elevated and two metabolites were downregulated in thermophile stress response to GVE2 infection. As an example, the upregulated L-norleucine was characterized. The data showed that L-norleucine had antiviral activity in thermophile. Furthermore, the in vitro and in vivo assays revealed that L-norleucine, as well as its derivative, significantly suppressed metastasis of gastric and breast cancer cells. L-norleucine interacted with hnRNPA2/B1 protein to inhibit the expressions of Twist1 and Snail, two inhibitors of E-cadherin, and promote the E-cadherin expression, leading to the inhibition of tumor metastasis. Therefore, our study presented that antiviral homeostasis-maintaining metabolites of microbes might be a promising source for anti-tumor drugs.
Sigma-Aldrich
Boc-Nle-OH, ≥99.0% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
L-Norvaline, arginase inhibitor
Pricing and availability is not currently available.