Merck
  • Harnessing the β-Silicon Effect for Regioselective and Stereoselective Rhodium(II)-Catalyzed C-H Functionalization by Donor/Acceptor Carbenes Derived from 1-Sulfonyl-1,2,3-triazoles.
Harnessing the β-Silicon Effect for Regioselective and Stereoselective Rhodium(II)-Catalyzed C-H Functionalization by Donor/Acceptor Carbenes Derived from 1-Sulfonyl-1,2,3-triazoles.
Organic letters (2018-04-12)
Zachary J Garlets, Huw M L Davies
ABSTRACT
The regioselective and enantioselective intermolecular sp3 C-H functionalization of silicon-substituted alkanes was accomplished using Rh2( S-NTTL)4 with readily available 1-sulfonyl-1,2,3-triazoles as carbene precursors. These reactions generate a diverse array of stereodefined substituted silaalkanes.
Sigma-Aldrich
Rh2(S-PTAD)4
Sigma-Aldrich
Rh2(R-PTAD)4
Sigma-Aldrich
Rh2(S-TCPTAD)4