Merck
  • Expression of neutrophil gelatinase-associated lipocalin (NGAL) in the gut in Crohn's disease.
Expression of neutrophil gelatinase-associated lipocalin (NGAL) in the gut in Crohn's disease.
Cell and tissue research (2018-06-06)
Silje Thorsvik, Ingunn Bakke, Atle van Beelen Granlund, Elin Synnøve Røyset, Jan Kristian Damås, Ann Elisabet Østvik, Arne Kristian Sandvik
ABSTRACT
The antimicrobial glycoprotein neutrophil gelatinase-associated lipocalin (NGAL) is strongly expressed in several infectious, inflammatory and malignant disorders, among these inflammatory bowel disease (IBD). Fecal and serum NGAL is elevated during active IBD and we have recently shown that fecal NGAL is a novel biomarker for IBD with a test performance comparable to the established fecal biomarker calprotectin. This study examines expression of NGAL in the healthy gut and in Crohn's disease (CD), with emphasis on the previously unexplored small intestine. Pinch biopsies were taken from active and inactive CD in jejunum, ileum and colon and from the same sites in healthy controls. Microarray gene expression showed that the NGAL gene, LCN2, was the second most upregulated among 1820 differentially expressed genes in terminal ileum comparing active CD and controls (FC 5.86, p = 0.027). Based on immunohistochemistry and in situ hybridization findings, this upregulation most likely represented increased expression in epithelial cells. Double immunofluorescence showed NGAL expression in 49% (range 19-70) of Paneth cells (PCs) in control ileum with no change during inflammation. In healthy jejunum, the NGAL expression in PCs was weak to none but markedly increased during active CD. We further found NGAL also in metaplastic PCs in colon. Finally, we show for the first time that NGAL is expressed in enteroendocrine cells in small intestine as well as in colon.
Sigma-Aldrich
Anti-DEFA6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution