Merck
  • Liver proteomic analysis of postpartum Holstein cows exposed to heat stress or cooling conditions during the dry period.
Liver proteomic analysis of postpartum Holstein cows exposed to heat stress or cooling conditions during the dry period.
Journal of dairy science (2017-11-13)
Amy L Skibiel, Maya Zachut, Bruno C do Amaral, Yishai Levin, Geoffrey E Dahl
ABSTRACT
Heat stress negatively affects cow performance, compromises immune function, and increases susceptibility to metabolic disorders, particularly during the dry period and as cows transition from gestation to lactation. Metabolic adaptations of the liver are critical for successful transition, yet it is unclear how heat stress affects metabolic pathways within the liver at the proteomic level. The objective of this study was to investigate the liver proteome of postpartum cows that were cooled or heat stressed during the dry period to gain insight into how protein expression is altered by prior heat stress and may contribute to performance and disease outcomes. During the dry period, cows were either housed in shaded barns with fans and water soakers [cooled group (CL); n = 5] or in shaded barns lacking these cooling devices [heat-stressed group (HT); n = 5]. Liver biopsies were collected at 2 d postpartum, and protein content was analyzed by label-free quantitative shotgun proteomics (nanoscale liquid chromatography coupled to tandem mass spectrometry). In the most comprehensive bovine liver proteomics analysis completed to date, we identified 3,270 proteins, 75 of which were differentially expressed between HT and CL cows (fold change ±1.2). The top pathways differing between HT and CL cows were oxidative phosphorylation, mitochondrial dysfunction, farnesoid X receptor/retinoid X receptor (FXR/RXR) activation, and the methylmalonyl pathway. Cooling cows during the dry period likely improves ATP production, reduces oxidative stress, and prevents excessive accumulation of hepatic triglycerides and cholesterol, which may contribute to greater milk yield and lower susceptibility to transition-related diseases.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Mouse IgG (Fab specific)–Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody