Merck
  • The Influence of Metal Ions on the Dynamics of Supported Phospholipid Langmuir Films.
The Influence of Metal Ions on the Dynamics of Supported Phospholipid Langmuir Films.
Langmuir : the ACS journal of surfaces and colloids (2017-03-08)
Stephen M Baumler, Gary J Blanchard
ABSTRACT
The translational diffusion dynamics of the zwitterionic lipid 1,2-dimyristoyl-sn-glycero-3-phosphocholine (DMPC) at a planar phosphorylated support surface containing metal ions (Mg2+, Ca2+, Ba2+, Ni2+, Zn2+, Cd2+, Zr4+) was investigated using X-ray photoelectron spectroscopy (XPS) and fluorescence recovery after photobleaching (FRAP). Fluorescence recovery curves yielded diffusion constants on the order of 2-5 μm2/s for the chromophore-tagged 12:0 NBD-Lyso-PC. Ionic interactions between the zwitterionic headgroup and metal ions were found to play a secondary role in mediating lipid fluidity. This work provides quantitative insight into the extent to which the fluidity of a supported lipid film is mediated by the ionic interactions between headgroup and surface versus that of the lipid-lipid tailgroup interactions.
Product Description
Avanti
12:0 Lyso NBD PC, Avanti Research - A Croda Brand 810128P, powder
Avanti
14:0 PC (DMPC), 1,2-dimyristoyl-sn-glycero-3-phosphocholine, powder