Merck
  • How Detergent Impacts Membrane Proteins: Atomic-Level Views of Mitochondrial Carriers in Dodecylphosphocholine.
How Detergent Impacts Membrane Proteins: Atomic-Level Views of Mitochondrial Carriers in Dodecylphosphocholine.
The journal of physical chemistry letters (2018-02-06)
Vilius Kurauskas, Audrey Hessel, Peixiang Ma, Paola Lunetti, Katharina Weinhäupl, Lionel Imbert, Bernhard Brutscher, Martin S King, Rémy Sounier, Vincenza Dolce, Edmund R S Kunji, Loredana Capobianco, Christophe Chipot, François Dehez, Beate Bersch, Paul Schanda
ABSTRACT
Characterizing the structure of membrane proteins (MPs) generally requires extraction from their native environment, most commonly with detergents. Yet, the physicochemical properties of detergent micelles and lipid bilayers differ markedly and could alter the structural organization of MPs, albeit without general rules. Dodecylphosphocholine (DPC) is the most widely used detergent for MP structure determination by NMR, but the physiological relevance of several prominent structures has been questioned, though indirectly, by other biophysical techniques, e.g., functional/thermostability assay (TSA) and molecular dynamics (MD) simulations. Here, we resolve unambiguously this controversy by probing the functional relevance of three different mitochondrial carriers (MCs) in DPC at the atomic level, using an exhaustive set of solution-NMR experiments, complemented by functional/TSA and MD data. Our results provide atomic-level insight into the structure, substrate interaction and dynamics of the detergent-membrane protein complexes and demonstrates cogently that, while high-resolution NMR signals can be obtained for MCs in DPC, they systematically correspond to nonfunctional states.
Product Description
Avanti
Heart CA, Avanti Research - A Croda Brand 840012P, powder
Avanti
18:1 Cardiolipin, Avanti Research - A Croda Brand