Merck
  • Global quantitative TPA-based proteomics of mouse brain structures reveals significant alterations in expression of proteins involved in neuronal plasticity during aging.
Global quantitative TPA-based proteomics of mouse brain structures reveals significant alterations in expression of proteins involved in neuronal plasticity during aging.
Aging (2018-07-22)
Przemysław Duda, Olga Wójcicka, Jacek R Wiśniewski, Dariusz Rakus
ABSTRACT
Aging is believed to be the result of alterations of protein expression and accumulation of changes in biomolecules. Although there are numerous reports demonstrating changes in protein expression in brain during aging, only few of them describe global changes at the protein level. Here, we present the deepest quantitative proteomic analysis of three brain regions, hippocampus, cortex and cerebellum, in mice aged 1 or 12 months, using the total protein approach technique. In all the brain regions, both in young and middle-aged animals, we quantitatively measured over 5,200 proteins. We found that although the total protein expression in middle-aged brain structures is practically unaffected by aging, there are significant differences between young and middle-aged mice in the expression of some receptors and signaling cascade proteins proven to be significant for learning and memory formation. Our analysis demonstrates that the hippocampus is the most variable structure during natural aging and that the first symptoms of weakening of neuronal plasticity may be observed on protein level in middle-aged animals.
Sigma-Aldrich
Anti-NMDAR2B antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, ascites fluid, clone HM-2
Sigma-Aldrich
Anti-GLUR1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-NMDAR2A antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-GLUR4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Fluoroshield, histology mounting medium