Merck
  • The complex fate in plasma of gadolinium incorporated into high-density lipoproteins used for magnetic imaging of atherosclerotic plaques.
The complex fate in plasma of gadolinium incorporated into high-density lipoproteins used for magnetic imaging of atherosclerotic plaques.
Bioconjugate chemistry (2013-04-27)
Alessandra Barazza, Courtney Blachford, Orli Even-Or, Victor A Joaquin, Karen C Briley-Saebo, Wei Chen, Xian-Cheng Jiang, Willem J M Mulder, David P Cormode, Zahi A Fayad, Edward A Fisher
ABSTRACT
We have previously reported enhancing the imaging of atherosclerotic plaques in mice using reconstituted high density lipoproteins (HDL) as nanocarriers for the MRI contrast agent gadolinium (Gd). This study focuses on the underlying mechanisms of Gd delivery to atherosclerotic plaques. HDL, LDL, and VLDL particles containing Gd chelated to phosphatidyl ethanolamine (DTPA-DMPE) and a lipidic fluorophore were used to demonstrate the transfer of Gd-phospholipids among plasma lipoproteins in vitro and in vivo. To determine the basis of this transfer, the roles of phospholipid transfer protein (PLTP) and lipoprotein lipase (LpL) in mediating the migration of Gd-DTPA-DMPE among lipoproteins were investigated. The results indicated that neither was an important factor, suggesting that spontaneous transfer of Gd-DTPA-DMPE was the most probable mechanism. Finally, two independent mouse models were used to quantify the relative contributions of HDL and LDL reconstituted with Gd-DTPA-DMPE to plaque imaging enhancement by MR. Both sets of results suggested that Gd-DTPA-DMPE originally associated with LDL was about twice as effective as that injected in the form of Gd-HDL, and that some of Gd-HDL's effectiveness in vivo is indirect through transfer of the imaging agent to LDL. In conclusion, the fate of Gd-DTPA-DMPE associated with a particular type of lipoprotein is complex, and includes its transfer to other lipoprotein species that are then cleared from the plasma into tissues.
Product Description
Avanti
14:0 PE-DTPA (Gd), 1,2-dimyristoyl-sn-glycero-3-phosphoethanolamine-N-diethylenetriaminepentaacetic acid (gadolinium salt), powder
Pricing and availability is not currently available.