Merck
  • Dendrite-Specific Amplification of Weak Synaptic Input during Network Activity In Vivo.
Dendrite-Specific Amplification of Weak Synaptic Input during Network Activity In Vivo.
Cell reports (2018-09-27)
Leiron Ferrarese, Jean-Sébastien Jouhanneau, Michiel W H Remme, Jens Kremkow, Gergely Katona, Balázs Rózsa, Susanne Schreiber, James F A Poulet
ABSTRACT
Excitatory synaptic input reaches the soma of a cortical excitatory pyramidal neuron via anatomically segregated apical and basal dendrites. In vivo, dendritic inputs are integrated during depolarized network activity, but how network activity affects apical and basal inputs is not understood. Using subcellular two-photon stimulation of Channelrhodopsin2-expressing layer 2/3 pyramidal neurons in somatosensory cortex, nucleus-specific thalamic optogenetic stimulation, and paired recordings, we show that slow, depolarized network activity amplifies small-amplitude synaptic inputs targeted to basal dendrites but reduces the amplitude of all inputs from apical dendrites and the cell soma. Intracellular pharmacology and mathematical modeling suggests that the amplification of weak basal inputs is mediated by postsynaptic voltage-gated channels. Thus, network activity dynamically reconfigures the relative somatic contribution of apical and basal inputs and could act to enhance the detectability of weak synaptic inputs.
Sigma-Aldrich
Phosphocreatine disodium salt hydrate, ≥97%
Sigma-Aldrich
Urethane, ≥99%
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Magnesium chloride hexahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Biocytin, ≥98% (TLC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Agarose, High EEO
Sigma-Aldrich
Sodium phosphate monobasic monohydrate, ACS reagent, ≥98%
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Calcium chloride dihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000