Merck
  • Generation of Tumor-Reactive T Cells by Co-culture of Peripheral Blood Lymphocytes and Tumor Organoids.
Generation of Tumor-Reactive T Cells by Co-culture of Peripheral Blood Lymphocytes and Tumor Organoids.
Cell (2018-08-14)
Krijn K Dijkstra, Chiara M Cattaneo, Fleur Weeber, Myriam Chalabi, Joris van de Haar, Lorenzo F Fanchi, Maarten Slagter, Daphne L van der Velden, Sovann Kaing, Sander Kelderman, Nienke van Rooij, Monique E van Leerdam, Annekatrien Depla, Egbert F Smit, Koen J Hartemink, Rosa de Groot, Monika C Wolkers, Norman Sachs, Petur Snaebjornsson, Kim Monkhorst, John Haanen, Hans Clevers, Ton N Schumacher, Emile E Voest
ABSTRACT
Cancer immunotherapies have shown substantial clinical activity for a subset of patients with epithelial cancers. Still, technological platforms to study cancer T-cell interactions for individual patients and understand determinants of responsiveness are presently lacking. Here, we establish and validate a platform to induce and analyze tumor-specific T cell responses to epithelial cancers in a personalized manner. We demonstrate that co-cultures of autologous tumor organoids and peripheral blood lymphocytes can be used to enrich tumor-reactive T cells from peripheral blood of patients with mismatch repair-deficient colorectal cancer and non-small-cell lung cancer. Furthermore, we demonstrate that these T cells can be used to assess the efficiency of killing of matched tumor organoids. This platform provides an unbiased strategy for the isolation of tumor-reactive T cells and provides a means by which to assess the sensitivity of tumor cells to T cell-mediated attack at the level of the individual patient.
Sigma-Aldrich
Nutlin-3, ≥98% (HPLC), powder
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Nutlin-3a, ≥98% (HPLC)
Sigma-Aldrich
Phorbol 12-Myristate 13-Acetate