Merck
Late metabolic precursors for selective aromatic residue labeling.
Journal of biomolecular NMR (2018-05-29)
Julia Schörghuber, Leonhard Geist, Gerald Platzer, Michael Feichtinger, Marilena Bisaccia, Lukas Scheibelberger, Frederik Weber, Robert Konrat, Roman J Lichtenecker
ABSTRACT
In recent years, we developed a toolbox of heavy isotope containing compounds, which serve as metabolic amino acid precursors in the E. coli-based overexpression of aromatic residue labeled proteins. Our labeling techniques show excellent results both in terms of selectivity and isotope incorporation levels. They are additionally distinguished by low sample production costs and meet the economic demands to further implement protein NMR spectroscopy as a routinely used method in drug development processes. Different isotopologues allow for the assembly of optimized protein samples, which fulfill the requirements of various NMR experiments to elucidate protein structures, analyze conformational dynamics, or probe interaction surfaces. In the present article, we want to summarize the precursors we developed so far and give examples of their special value in the probing of protein-ligand interaction.
Sigma-Aldrich
D-(−)-Erythrose, ≥75% (TLC), syrup
Pricing and availability is not currently available.