Merck
Hyperspectral Canopy Sensing of Wheat Septoria Tritici Blotch Disease.
Frontiers in plant science (2018-09-04)
Kang Yu, Jonas Anderegg, Alexey Mikaberidze, Petteri Karisto, Fabio Mascher, Bruce A McDonald, Achim Walter, Andreas Hund
ABSTRACT
Producing quantitative and reliable measures of crop disease is essential for resistance breeding, but is challenging and time consuming using traditional phenotyping methods. Hyperspectral remote sensing has shown potential for the detection of plant diseases, but its utility for phenotyping large and diverse populations of plants under field conditions requires further evaluation. In this study, we collected canopy hyperspectral data from 335 wheat varieties using a spectroradiometer, and we investigated the use of canopy reflectance for detecting the Septoria tritici blotch (STB) disease and for quantifying the severity of infection. Canopy- and leaf-level infection metrics of STB based on traditional visual assessments and automated analyses of leaf images were used as ground truth data. Results showed (i) that canopy reflectance and the selected spectral indices show promise for quantifying STB infections, and (ii) that the normalized difference water index (NDWI) showed the best performance in detecting STB compared to other spectral indices. Moreover, partial least squares (PLS) regression models allowed for an improvement in the prediction of STB metrics. The PLS discriminant analysis (PLSDA) model calibrated based on the spectral data of four reference varieties was able to discriminate between the diseased and healthy canopies among the 335 varieties with an accuracy of 93% (Kappa = 0.60). Finally, the PLSDA model predictions allowed for the identification of wheat genotypes that are potentially more susceptible to STB, which was confirmed by the STB visual assessment. This study demonstrates the great potential of using canopy hyperspectral remote sensing to improve foliar disease assessment and to facilitate plant breeding for disease resistance.
Product Description
Supelco
Prothioconazole, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Bixafen, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Supelco
Spiroxamine, PESTANAL®, analytical standard
Pricing and availability is not currently available.