Merck
  • Quorum sensing system and influence on food spoilage in Pseudomonas fluorescens from turbot.
Quorum sensing system and influence on food spoilage in Pseudomonas fluorescens from turbot.
Journal of food science and technology (2018-08-02)
Tingting Li, Bing Yang, Xuepeng Li, Jianrong Li, Guohua Zhao, Jianquan Kan
ABSTRACT
The spoilage of aquatic products is mainly caused by the bacterial growth, and the specific spoilage organism (SSO) plays an important role. Quorum sensing (QS) is a microbial cell-cell communication system which is coordinated with the population density, and is controlled by N-acyl-homoserine lactone (AHLs) as the Gram-negative bacteria communication signals. In this study, the SSO was Pseudomonas fluorescens (PF-04), isolated from the turbot (Scophthalmus maximus L.) during aerobically refrigerated storage. The supernatant extract of PF-04 tested the AHLs activities utilizing biosensor Chromobacterium violaceum CV026. AHL production was influenced by the environment temperature, and AHL production reduced obviously at 10 °C compare with 25 °C. In Luria-Bertani (LB) supplemented with 0.5-1.0% NaCl, AHL production reached the maximum. The AHL production was also regulated by pH of culture medium, acidic condition was conducive to persistent existence of the AHL molecules, but the alkaline environment would cause chemically unstable of AHL molecules. QS system in P. flurosecens played an imperative role in biofilm formation, protease and siderophore production. AHLs could regulate above three factors in PF-04. In summary, this study showed that (1) the influence of different environmental conditions (temperature, NaCl and pH) on AHL production revealed the correlation of QS in foods and (2) that proved the effect of external AHLs to regulate the biofilm formation, protease and siderophore production in PF-04.
Sigma-Aldrich
N-Decanoyl-DL-homoserine lactone, ≥97.0% (HPLC)
Pricing and availability is not currently available.