Merck
  • Identification of non volatile migrant compounds and NIAS in polypropylene films used as food packaging characterized by UPLC-MS/QTOF.
Identification of non volatile migrant compounds and NIAS in polypropylene films used as food packaging characterized by UPLC-MS/QTOF.
Talanta (2018-07-22)
Paula Vera, Elena Canellas, Cristina Nerín
ABSTRACT
Migration of non volatile compounds from twenty six PP films used as food contact materials has been studied in four simulants (ethanol 95% and 10%, acetic acid 3% and Tenax ®) and analyzed by UPLC-MS/QTOF. Seventy six compounds have been identified, where 76% of them were non-intentionally added substances (NIAS) coming from degradation of additives used, such as methyl or ethyl or hexyl-3-(3,5-di-tert-butyl-4-hydroxyphenyl) from irganox 1076 and irganox 1010 degradation; or impurities such as N,N-bis(2-hydroxyethyl) amines, or compounds of unknown origin, like hydro-ceramides. The most common compounds found were glyceryl monostearate or monopalmitate, erucamide, irganox 1010, irgafos 168, irgafos 168 OXO, N,N-bis(2-hydroxyethyl) tridecylamine and N,N-bis(2-hydroxyethyl) pentadecylamine. Six films didn't comply with the European Regulation Nº 10/2011/EU, where irganox 1010 and the group of N,N-bis(2-hydroxyethyl) amines exceeded their SMLs. Other films surpassed the maximum concentration recommended by Cramer for the compounds of class II (degradation products) or III (amide compounds) when ethanol 95% was used as simulant.
Sigma-Aldrich
cis-13-Docosenoamide
Sigma-Aldrich
3,5-Ditert-butyl-4-hydroxybenzaldehyde, AldrichCPR
Sigma-Aldrich
4-sec-Butyl-2,6-di-tert-butylphenol, 96%